Dimethyl terephthalate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Terephthalic acid dimethyl ester
M-Clarity™ quality level = MQ100
86440-1KG
86440-250G
86440-50G
SIAL86440-1KGEA
69.9
GBP
SIAL86440-1KG
SIAL86440-250G
SIAL86440-50G
Dimethyl terephthalate, Sigma-Aldrich®
Dimethyl terephthalate
Formula:
C₁₀H₁₀O₄ MW: 194.19 g/mol Boiling Pt: 288 °C (1013 hPa) Melting Pt: 139 °C Density: 1.2 g/cm³ (20 °C) Flash Pt: 141 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008440 CAS Number: 120-61-6 EINECS: 204-411-8 Merck Index: 12,09306 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...