Dimethyl terephthalate ≥99%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Terephthalic acid dimethyl ester

A13220.0E A13220.36
A13220.0EEA 0 GBP
A13220.0E A13220.36
Dimethyl terephthalate ≥99%
Dimethyl terephthalate

Purity: 99% CAS No.: 120-61-6 Molecular Formula: C10H10O4 Formula Weight: 194.19 Flash Point: 146°(294°F) Melting Point: 140-143° Boiling Point: 288° Beilstein Reference: 1107185 Merck Reference: 14,9162 MDL No.: MFCD00008440

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Boiling Pt: 288 °C
Melting Pt: 140…143 °C
Density: 1.290
Flash Pt: 146 °C (294 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008440
CAS Number: 120-61-6
EINECS: 204-411-8
Merck Index: 14,9162

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR