Diethyl phthalate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Phthalic acid diethyl ester

MQ200

524972-100ML 524972-1L 524972-20L 524972-5ML
SIAL524972-100MLEA 38.9 GBP
SIAL524972-100ML SIAL524972-1L SIAL524972-20L SIAL524972-5ML
Diethyl phthalate, Sigma-Aldrich®
Diethyl phthalate
Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
Boiling Pt: 298
Melting Pt: –3 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 156 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00009111
CAS Number: 84-66-2
EINECS: 201-550-6
Merck Index: 12,07527
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d0a6c6b14ead75148c0661141b5e8d00e6fc1a9&vid=2461e9f0e230ca44b5372d1233410fbf47a564c7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR