Methyl-4-hydroxybenzoate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ200

H6654-100G H6654-1KG
SIALH6654-100GEA 118 GBP
SIALH6654-100G SIALH6654-1KG
Methyl-4-hydroxybenzoate, Sigma-Aldrich®
Methyl 4-hydroxybenzoate
Formula: HOC₆H₄CO₂CH₃
Boiling Pt: 270…280 °C (1013 hPa)
Melting Pt: 125…128 °C
Density: 1.1208 g/cm³ (137 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002352
CAS Number: 99-76-3
EINECS: 202-785-7
Merck Index: 13,06129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84321052052f4be2a53d8d7ebac7fabe6fc5a435&vid=ede765fad275b2515a8c57ba4499bff313a18983&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR