Methyl-2-isothiocyanatobenzoate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M1368-C.5 M1368-C.25
XXXXM1368-C.5EA 0 GBP
XXXXM1368-C.5 XXXXM1368-C.25
Methyl-2-isothiocyanatobenzoate
Methyl 2-isothiocyanatobenzoate
Formula: C₉H₇NO₂S
MW: 193.23 g/mol
Boiling Pt: 104 °C (1013 hPa)
Density: 1.25 g/cm³ (20 °C)
MDL Number: MFCD00041062
CAS Number: 16024-82-1
EINECS: 240-164-2
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR