Benzanilide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N-Phenylbenzamide
M-Clarity™ quality level = MQ100
108227-100G
108227-5G
SIAL108227-100GEA
88.8
GBP
SIAL108227-100G
SIAL108227-5G
Benzanilide, Sigma-Aldrich®
Benzanilide
Formula:
C₁₃H₁₁NO MW: 197.24 g/mol Boiling Pt: 247 °C (1013 hPa) Melting Pt: 162…165 °C Density: 1.32 g/cm³ (20 °C) |
MDL Number:
MFCD00003069 CAS Number: 93-98-1 EINECS: 202-292-7 Merck Index: 13,01060 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...