4-Hydroxybenzhydrazide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Hydroxybenzoic hydrazide, 4-Hydroxybenzohydrazide, p-Hydroxybenzoic acid hydrazide, 4-Hydroxybenzene-1-carbohydrazide, 4-Hydroxybenzoic acid hydrazide

018715-10G 018715-25G
FLUO018715-10GEA 0 GBP
FLUO018715-10G FLUO018715-25G
4-Hydroxybenzhydrazide
4-Hydroxybenzhydrazide
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 370
Melting Pt: 266…269 °C
Storage Temperature: Ambient
MDL Number: MFCD00007605
CAS Number: 5351-23-5
EINECS: 226-326-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR