Salicylhydrazide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Hydroxybenzoic acid hydrazide, Salicylohydrazide, 2-Hydroxybenzene-1-carbohydrazide, Salicylic acid hydrazide, 2-Hydroxybenzohydrazide, 2-Hydroxybenzhydrazide, 2-Hydroxybenzoic hydrazide

018713-5G 018713-25G
FLUO018713-5GEA 0 GBP
FLUO018713-5G FLUO018713-25G
Salicylhydrazide
Salicylhydrazide
Formula: C₇H₈N₂O₂
Melting Pt: 290…291 °C
Density: 1,424 g/cm³ (20 °C)
MDL Number: MFCD00007599
CAS Number: 936-02-7
EINECS: 213-311-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR