2,5-Dimethoxybenzhydrazide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,5-Dimethoxybenzoic acid hydrazide, 2,5-Dimethoxybenzoic hydrazide, 2,5-Dimethoxybenzene-1-carbohydrazide, 2,5-Dimethoxybenzohydrazide

018569-1G 018569-5G
FLUO018569-1GEA 0 GBP
FLUO018569-1G FLUO018569-5G
2,5-Dimethoxybenzhydrazide
2,5-Dimethoxybenzhydrazide
Formula: C₉H₁₂N₂O₃
MW: 196.21 g/mol
MDL Number: MFCD00017049
CAS Number: 17894-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR