p-Anisohydrazide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Methoxybenzoic acid hydrazide, 4-Methoxybenzoic hydrazide, 4-Methoxybenzhydrazide, 4-Methoxybenzohydrazide, 4-Methoxybenzene-1-carbohydrazide, 4-Anisohydrazide

558346-25G 558346-5G
SIAL558346-25GEA 77.3 GBP
SIAL558346-25G SIAL558346-5G
p-Anisohydrazide, Sigma-Aldrich®
p-Anisohydrazide
Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD00017073
CAS Number: 3290-99-1
EINECS: 221-952-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR