Salicylamide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Hydroxybenzamide
M-Clarity™ quality level = MQ100
84230-1KG
84230-250G
84230-50G
SIAL84230-1KGEA
135
GBP
SIAL84230-1KG
SIAL84230-250G
SIAL84230-50G
Salicylamide, Sigma-Aldrich®
Salicylamide
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Boiling Pt: 318 Melting Pt: 137 Density: 1.33 g/cm³ (20 °C) |
MDL Number:
MFCD00007978 CAS Number: 65-45-2 EINECS: 200-609-3 Merck Index: 12,08480 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...