o-Acetoxybenzoyl chloride ≥97%
Supplier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
2-(Chlorocarbonyl)phenyl acetate, O-Acetylsalicyloyl chloride, 2-Acetylsalicyloyl chloride, 2-Acetoxybenzoyl chloride
Purity: 97%
CAS No.: 5538-51-2
Molecular Formula: C9H7ClO3
Formula Weight: 198.61
Flash Point:
Formula:
C₉H₇ClO₃ Boiling Pt: 134…136 °C (12 mmHg) Melting Pt: 47…50 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000663 CAS Number: 5538-51-2 EINECS: 226-899-1 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...