3-Hydroxybenzonitrile, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
m-Hydroxybenzonitrile, m-Cyanophenol, 3-Cyanophenol
M-Clarity™ quality level = MQ100
C93800-10G
C93800-5G
SIALC93800-10GEA
149
GBP
SIALC93800-10G
SIALC93800-5G
3-Hydroxybenzonitrile, Sigma-Aldrich®
3-Hydroxybenzonitrile
Formula:
C₇H₅NO MW: 119.12 g/mol Boiling Pt: 265 °C (1013 hPa) Melting Pt: 84.5…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002252 CAS Number: 873-62-1 EINECS: 212-845-4 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...