2,3-Dimethoxybenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

023875-25G 023875-5G
FLUO023875-25GEA 0 GBP
FLUO023875-25G FLUO023875-5G
2,3-Dimethoxybenzonitrile
2,3-Dimethoxybenzonitrile
Formula: C₉H₉NO₂
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 47 °C
MDL Number: MFCD00001784
CAS Number: 5653-62-3
EINECS: 227-097-4
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR