2-Cyanophenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008779-5G
008779-1G
FLUO008779-5GEA
0
GBP
FLUO008779-5G
FLUO008779-1G
2-Cyanophenyl isothiocyanate
2-Cyanophenyl isothiocyanate
Formula:
C₈H₄N₂S MW: 160.2 g/mol |
MDL Number:
MFCD00060361 CAS Number: 81431-98-3 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...