2,6-Dichlorobenzenesulphonyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,6-Dichlorobenzene-1-sulphonyl chloride
CFF419.10
CFF419.25
CFF419.50
003583-5G
003583-25G
CHMPCFF419.10EA
0
GBP
CHMPCFF419.10
CHMPCFF419.25
CHMPCFF419.50
FLUO003583-5G
FLUO003583-25G
2,6-Dichlorobenzenesulphonyl chloride
2,6-Dichlorobenzenesulphonyl chloride
Formula:
C₆H₃Cl₃O₂S Melting Pt: 54…56 °C |
MDL Number:
MFCD00052311 CAS Number: 6579-54-0 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...