2,4,6-Trichlorobenzenesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013069-5G 013069-25G
FLUO013069-5GEA 0 GBP
FLUO013069-5G FLUO013069-25G
2,4,6-Trichlorobenzenesulphonyl chloride
2,4,6-Trichlorobenzenesulphonyl chloride
Formula: C₆H₂Cl₄O₂S
Melting Pt: 49 °C
MDL Number: MFCD00052973
CAS Number: 51527-73-2
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR