4-Chlorophenyl phenyl ether

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Chlorodiphenyl ether, 1-Chloro-4-phenoxybenzene

002531-5G 002531-25G
FLUO002531-5GEA 0 GBP
FLUO002531-5G FLUO002531-25G
4-Chlorophenyl phenyl ether
4-Chlorophenyl phenyl ether
Formula: C₁₂H₉ClO
MW: 204.66 g/mol
Boiling Pt: 295
Melting Pt: –8 °C
Density: 1.193 g/cm³ (20 °C)
MDL Number: MFCD00055431
CAS Number: 7005-72-3
EINECS: 230-281-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR