1-Chloro-3,5-dimethoxybenzene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Chloro-1,3-dimethoxybenzene

M-Clarity™ quality level = MQ100

209252-25G 209252-5G
SIAL209252-25GEA 206 GBP
SIAL209252-25G SIAL209252-5G
1-Chloro-3,5-dimethoxybenzene, Sigma-Aldrich®
1-Chloro-3,5-dimethoxybenzene
Formula: C₈H₉ClO₂
MW: 172.61 g/mol
Boiling Pt: 234 °C (1013 hPa)
Melting Pt: 38 °C
MDL Number: MFCD00008382
CAS Number: 7051-16-3
EINECS: 230-330-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR