5-Chloro-2-nitroanisole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

SIALC58606-10GEA 82.6 GBP
SIALC58606-10G SIALC58606-25G
5-Chloro-2-nitroanisole, Sigma-Aldrich®
5-Chloro-2-nitroanisole
Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
Melting Pt: 71 °C
MDL Number: MFCD00007288
CAS Number: 6627-53-8
EINECS: 229-594-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR