2-Chloro-5-nitrophenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008771-5G
008771-1G
FLUO008771-5GEA
0
GBP
FLUO008771-5G
FLUO008771-1G
2-Chloro-5-nitrophenyl isothiocyanate
2-Chloro-5-nitrophenyl isothiocyanate
Formula:
C₇H₃ClN₂O₂S |
MDL Number:
MFCD00060632 CAS Number: 57135-68-9 EINECS: 260-586-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...