4-Chlorophenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Chlorophenylisothiocyanate, 1-Chloro-4-isothiocyanatobenzene
008775-5G
008775-25G
008775-10G
FLUO008775-5GEA
0
GBP
FLUO008775-5G
FLUO008775-25G
FLUO008775-10G
4-Chlorophenyl isothiocyanate
4-Chlorophenyl isothiocyanate
Formula:
C₇H₄ClNS MW: 169.63 g/mol Boiling Pt: 135…142 °C (22 mmHg) Melting Pt: 46 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004810 CAS Number: 2131-55-7 EINECS: 218-358-3 UN: 2923 ADR: 8,III |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...