3-Chlorophenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008774-5G
008774-25G
008774-10G
FLUO008774-5GEA
0
GBP
FLUO008774-5G
FLUO008774-25G
FLUO008774-10G
3-Chlorophenyl isothiocyanate
3-Chlorophenyl isothiocyanate
Formula:
C₇H₄ClNS MW: 169.63 g/mol Boiling Pt: 64…65 °C (0.4 mmHg) Melting Pt: 45 °C Density: 1.281 g/cm³ (20 °C) |
MDL Number:
MFCD00004805 CAS Number: 2392-68-9 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...