2,6-Dichlorophenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
008814-1G
008814-5G
008814-25G
FLUO008814-1GEA
0
GBP
FLUO008814-1G
FLUO008814-5G
FLUO008814-25G
2,6-Dichlorophenyl isothiocyanate
2,6-Dichlorophenyl isothiocyanate
Formula:
C₇H₃Cl₂NS Boiling Pt: 100 °C (0.3 mmHg) Melting Pt: 44 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00041056 CAS Number: 6590-95-0 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...