2-Chloro-5-iodobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009093-5G 009093-25G 009093-100G
FLUO009093-5GEA 0 GBP
FLUO009093-5G FLUO009093-25G FLUO009093-100G
2-Chloro-5-iodobenzoic acid
2-Chloro-5-iodobenzoic acid
Formula: C₇H₄ClIO₂
MDL Number: MFCD00079731
CAS Number: 19094-56-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR