3-Chloro-2-fluorobenzenesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

021393-5G 021393-25G
FLUO021393-5GEA 0 GBP
FLUO021393-5G FLUO021393-25G
3-Chloro-2-fluorobenzenesulphonyl chloride
3-Chloro-2-fluorobenzenesulphonyl chloride
Formula: C₆H₃Cl₂FO₂S
MDL Number: MFCD03094234
CAS Number: 351003-48-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR