2-Chloro-3,6-difluoroanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005584-1G 005584-5G
FLUO005584-1GEA 0 GBP
FLUO005584-1G FLUO005584-5G
2-Chloro-3,6-difluoroanisole
2-Chloro-3,6-difluoroanisole
Formula: C₇H₅ClF₂O
MDL Number: MFCD01631451
CAS Number: 261762-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR