2-Chloro-1-fluoro-4-nitrobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002152-100G 002152-500G 002152-50G
FLUO002152-100GEA 0 GBP
FLUO002152-100G FLUO002152-500G FLUO002152-50G
2-Chloro-1-fluoro-4-nitrobenzene
2-Chloro-1-fluoro-4-nitrobenzene
Formula: C₆H₃ClFNO₂
Boiling Pt: 227…228 °C (1013 hPa)
Melting Pt: 41.5 °C
MDL Number: MFCD00007206
CAS Number: 350-30-1
EINECS: 206-499-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR