1-Chloro-2-fluoro-5-iodobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-1-fluoro-4-iodobenzene, 3-Chloro-4-fluoroiodobenzene

004453-25G 004453-100G
FLUO004453-25GEA 0 GBP
FLUO004453-25G FLUO004453-100G
1-Chloro-2-fluoro-5-iodobenzene
1-Chloro-2-fluoro-5-iodobenzene
Formula: C₆H₃ClFI
MW: 256.45 g/mol
MDL Number: MFCD00042210
CAS Number: 156150-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR