4-Chloro-3-fluorobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
005183-1G
005183-5G
005183-25G
FLUO005183-1GEA
0
GBP
FLUO005183-1G
FLUO005183-5G
FLUO005183-25G
4-Chloro-3-fluorobenzoic acid
4-Chloro-3-fluorobenzoic acid
Formula:
C₇H₄ClFO₂ |
MDL Number:
MFCD00143290 CAS Number: 403-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...