3-Chloro-5-fluorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

CFF934.1 005181-1G 005181-25G 005181-5G
CHMPCFF934.1EA 0 GBP
CHMPCFF934.1 FLUO005181-1G FLUO005181-25G FLUO005181-5G
3-Chloro-5-fluorobenzoic acid
3-Chloro-5-fluorobenzoic acid
Formula: C₇H₄ClFO₂
MDL Number: MFCD00973904
CAS Number: 25026-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR