3-Chloro-2,6-difluorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

005592-5G 005592-10G 005592-1G
FLUO005592-5GEA 0 GBP
FLUO005592-5G FLUO005592-10G FLUO005592-1G
3-Chloro-2,6-difluorobenzoic acid
3-Chloro-2,6-difluorobenzoic acid
Formula: C₇H₃ClF₂O₂
MW: 192.55 g/mol
MDL Number: MFCD01631322
CAS Number: 225104-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR