2-Chloro-5-fluorobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
008647-1G
008647-5G
008647-25G
FLUO008647-1GEA
0
GBP
FLUO008647-1G
FLUO008647-5G
FLUO008647-25G
2-Chloro-5-fluorobenzoic acid
2-Chloro-5-fluorobenzoic acid
Formula:
C₇H₄ClFO₂ |
MDL Number:
MFCD00156967 CAS Number: 2252-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...