2-Chloro-3-fluorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF929.1 005185-1G 005185-5G
CHMPCFF929.1EA 0 GBP
CHMPCFF929.1 FLUO005185-1G FLUO005185-5G
2-Chloro-3-fluorobenzoic acid
2-Chloro-3-fluorobenzoic acid
Formula: C₇H₄ClFO₂
MDL Number: MFCD00973903
CAS Number: 102940-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR