2-Chloro-3,6-difluorobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

005590-5G 005590-10G 005590-25G
FLUO005590-5GEA 0 GBP
FLUO005590-5G FLUO005590-10G FLUO005590-25G
2-Chloro-3,6-difluorobenzoic acid
2-Chloro-3,6-difluorobenzoic acid
Formula: C₇H₃ClF₂O₂
MW: 192.55 g/mol
MDL Number: MFCD01631441
CAS Number: 287172-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR