3-Chloro-2,6-difluorobenzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005595-5G 005595-10G
FLUO005595-5GEA 0 GBP
FLUO005595-5G FLUO005595-10G
3-Chloro-2,6-difluorobenzoyl chloride
3-Chloro-2,6-difluorobenzoyl chloride
Formula: C₇H₂Cl₂F₂O
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 70 °C
MDL Number: MFCD01631324
CAS Number: 261762-43-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR