2-Chloro-3,6-difluorobenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

005586-5G 005586-10G 005586-1G
FLUO005586-5GEA 0 GBP
FLUO005586-5G FLUO005586-10G FLUO005586-1G
2-Chloro-3,6-difluorobenzaldehyde
2-Chloro-3,6-difluorobenzaldehyde
Formula: C₇H₃ClF₂O
MDL Number: MFCD01631443
CAS Number: 261762-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR