2-Chloro-5-nitrobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ100
125113-100G
125113-500G
SIAL125113-100GEA
60
GBP
SIAL125113-100G
SIAL125113-500G
2-Chloro-5-nitrobenzoic acid, Sigma-Aldrich®
2-Chloro-5-nitrobenzoic acid
Formula:
C₇H₄ClNO₄ MW: 201.57 g/mol Melting Pt: 164…167 °C Density: 0.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007294 CAS Number: 2516-96-3 EINECS: 219-739-7 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...