2-Amino-4-chlorobenzonitrile, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 5-Chloro-2-cyanoaniline

M-Clarity™ quality level = MQ100

405280-10G 405280-50G
SIAL405280-10GEA 111 GBP
SIAL405280-10G SIAL405280-50G
2-Amino-4-chlorobenzonitrile, Sigma-Aldrich®
2-Amino-4-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
MDL Number: MFCD00035927
CAS Number: 38487-86-4
EINECS: 253-967-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR