5-Chlorosalicylaldehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ100

SIAL447706-25GEA 61 GBP
SIAL447706-25G SIAL447706-5G
5-Chlorosalicylaldehyde, Sigma-Aldrich®
5-Chlorosalicylaldehyde
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 105 °C (12 mmHg)
Melting Pt: 100.3 °C
MDL Number: MFCD00003331
CAS Number: 635-93-8
EINECS: 211-244-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR