2-Chlorophenylboronic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (2-Chlorophenyl)metaboric acid, Dihydroxy(2-chlorophenyl)borane, (2-Chlorophenyl)boronic acid, 2-Chlorobenzeneboronic acid

M-Clarity™ quality level = MQ100

445215-1G 445215-5G
SIAL445215-1GEA 56 GBP
SIAL445215-1G SIAL445215-5G
2-Chlorophenylboronic acid, Sigma-Aldrich®
2-Chlorophenylboronic acid
Formula: C₆H₆BClO₂
MW: 156.38 g/mol
MDL Number: MFCD00674012
CAS Number: 3900-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR