2,3,4-Trimethoxybenzaldehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

152099-10G 152099-50G
SIAL152099-10GEA 41.8 GBP
SIAL152099-10G SIAL152099-50G
2,3,4-Trimethoxybenzaldehyde, Sigma-Aldrich®
2,3,4-Trimethoxybenzaldehyde
Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Boiling Pt: 312 °C (1013 hPa)
Melting Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00003310
CAS Number: 2103-57-3
EINECS: 218-271-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR