2-Nitrobenzaldehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

N10802-100G N10802-25G N10802-2G
SIALN10802-100GEA 149 GBP
SIALN10802-100G SIALN10802-25G SIALN10802-2G
2-Nitrobenzaldehyde, Sigma-Aldrich®
2-Nitrobenzaldehyde
Formula: C₇H₅NO₃
MW: 151.12 g/mol
Boiling Pt: 285
Melting Pt: 41…43 °C
Density: 1.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007132
CAS Number: 552-89-6
EINECS: 209-025-3
Merck Index: 12,06684

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR