3,4-Dibromobenzenesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018498-5G 018498-1G
FLUO018498-5GEA 0 GBP
FLUO018498-5G FLUO018498-1G
3,4-Dibromobenzenesulphonyl chloride
3,4-Dibromobenzenesulphonyl chloride
Formula: C₆H₃Br₂ClO₂S
MDL Number: MFCD00060709
CAS Number: 81903-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR