2,5-Dibromobenzenesulphonyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,5-Dibromobenzene-1-sulphonyl chloride
018497-5G
18497.14
018497-25G
FLUO018497-5GEA
0
GBP
FLUO018497-5G
FLUO18497.14
FLUO018497-25G
2,5-Dibromobenzenesulphonyl chloride
2,5-Dibromobenzenesulphonyl chloride
Formula:
C₆H₃Br₂ClO₂S MW: 334.42 g/mol |
MDL Number:
MFCD00060685 CAS Number: 23886-64-8 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...