4-Bromothioanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

003696-100G 003696-500G 003696-25G
FLUO003696-100GEA 0 GBP
FLUO003696-100G FLUO003696-500G FLUO003696-25G
4-Bromothioanisole
4-Bromothioanisole
Formula: C₇H₇BrS
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 35 °C
Density: 1.4584 g/cm³ (39 °C)
MDL Number: MFCD00000102
CAS Number: 104-95-0
EINECS: 203-255-8
UN: 3335
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR