2-Bromothioanisole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001732-5G
001732-25G
001732-100G
FLUO001732-5GEA
0
GBP
FLUO001732-5G
FLUO001732-25G
FLUO001732-100G
2-Bromothioanisole
2-Bromothioanisole
Formula:
C₇H₇BrS Boiling Pt: 67…69 °C (0.5 mmHg) Melting Pt: –24.5 °C Density: 1.5135 g/cm³ (16 °C) |
MDL Number:
MFCD00000066 CAS Number: 19614-16-5 EINECS: 243-183-4 UN: 3334 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...