5-Bromo-1,2,3-trimethoxybenzene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Bromo-3,4,5-trimethoxybenzene

M-Clarity™ quality level = MQ100

634530-100G 634530-10G
SIAL634530-100GEA 443 GBP
SIAL634530-100G SIAL634530-10G
5-Bromo-1,2,3-trimethoxybenzene, Sigma-Aldrich®
5-Bromo-1,2,3-trimethoxybenzene
Formula: C₉H₁₁BrO₃
MW: 247.09 g/mol
MDL Number: MFCD00017169
CAS Number: 2675-79-8
EINECS: 220-223-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR