4-Bromophenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001275-5G 001275-25G
FLUO001275-5GEA 0 GBP
FLUO001275-5G FLUO001275-25G
4-Bromophenyl isothiocyanate
4-Bromophenyl isothiocyanate
Formula: C₇H₄BrNS
Boiling Pt: 142…144 °C (14 mmHg)
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00004808
CAS Number: 1985-12-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR