2-Bromo-4,6-difluorophenol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-Bromo-3,5-difluoro-2-hydroxybenzene
013197-25G
013197-5G
FLUO013197-25GEA
0
GBP
FLUO013197-25G
FLUO013197-5G
2-Bromo-4,6-difluorophenol
2-Bromo-4,6-difluorophenol
Formula:
C₆H₃BrF₂O MW: 208.99 g/mol |
MDL Number:
MFCD00070752 CAS Number: 98130-56-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...